missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human SMARCAD1 GST (N-Term) Protein

Product Code. 18309679 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18309679 25 μg 25µg
18327279 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18309679 Supplier Novus Biologicals™ Supplier No. H00056916P0125ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 56916
Molecular Weight (g/mol) 144 kDa
Quantity 25 μg
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias ATP-dependent helicase 1, DKFZP762K2015, EC 3.6.1, EC 3.6.4.12, HEL1, hHEL1, KIAA1122ETL1DKFZp762K2015, subfamily a, containing DEAD/H box 1, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A containing DEAD/H box 1, SWI/SNF-related, matrix-associated actin-dependent regulator of chromatin
Gene Symbol SMARCAD1
Research Category DNA Repair
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein SMARCAD1
Show More Show Less
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.