missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SMAD4 Protein
A cDNA sequence encoding the SMAD4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13526-50ug
Additional Details : Weight : 0.01000kg
Specifications
SMAD4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MDNMSITNTP TSNDACLSIV HSLMCHRQGG ESETFAKRAI ESLVKKLKEK KDELDSLITA ITTNGAHPSK CVTIQRTLDG RLQVAGRKGF PHVIYARLWR WPDLHKNELK HVKYCQYAFD LKCDSVCVNP YHYERVVSPG IDLSGLTLQS NAPSSMMVKD EYVHDFEGQP SLSTEGHSIQ TIQHPPSNRA STETYSTPAL LAPSESNATS TANFPNIPVA STSQPASILG GSHSEGLLQI ASGPQPGQQQ NGFTGQPATY HHNSTTTWTG SRTAPYTPNL PHHQNGHLQH HPPMPPHPGH YWPVHNELAF QPPISNHPAP EYWCSIAYFE MDVQVGETFK VPSSCPIVTV DGYVDPSGGD RFCLGQLSNV HRTEAIERAR LHIGKGVQLE CKGEGDVWVR CLSDHAVFVQ SYYLDREAGR APGDAVHKIY PSAYIKVFDL RQCHRQMQQQ AATAQAAAAA QAAAVAGNIP GPGSVGGIAP AISLSAAAGI GVDDLRRLCI LRMSFVKGWG PDYPRQSIKE TPCWIEIHLH RALQLLDEVL HTMPIADPQP LD | |
Greater than 90.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SMAD4 protein solution contains 20mM Tris-HCl pH 8.0, and 20% glycerol. |