missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Slit3 (aa 1120-1523) Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
The Recombinant Human Slit3 (aa 1120-1523) Protein is derived from HEK293. The Recombinant Human Slit3 (aa 1120-1523) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | O75094 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 6586 |
| Molecular Weight (g/mol) | Observed M.W.: 56-63kDa, reducing conditions, Theoretical M.W.: 45kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell,HEK293-derived human Slit3 protein Thr1120-Ser1523,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Gene Alias | KIAA0814, MEGF5, MEGF5Multiple epidermal growth factor-like domains protein 5, Multiple EGF-like domains protein 5, SLIL2, SLIL2FLJ10764, slit homolog 3 (Drosophila), slit homolog 3 protein, SLIT1, slit2, Slit-3slit (Drosophila) homolog 3 |
| Common Name | Slit3 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction