missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Skp1 Protein

Product Code. 18242690 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
500 μg
Unit Size:
0.1mg
500µg
This item is not returnable. View return policy

Product Code. 18242690

Brand: Novus Biologicals™ NBP130173

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

An un-tagged recombinant protein corresponding to the amino acids 1- 160 of Human Skp1 The Recombinant Human Skp1 Protein is derived from E. coli. The Recombinant Human Skp1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_008861
For Use With (Application) ELISA, Peptide ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 50mM NaCl, 10% glycerol
Gene ID (Entrez) 6500
Molecular Weight (g/mol) 18kDa
Name Skp1 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen 1- 160aa, Human
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.