missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Sirtuin 1/SIRT1 Protein

Product Code. 18389037
Click to view available options
:
50ug; Carrier Free
This item is not returnable. View return policy

Product Code. 18389037

Brand: R&D Systems™ 7714DA050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

The Recombinant Human Sirtuin 1/SIRT1 Protein is derived from E. coli. The Recombinant Human Sirtuin 1/SIRT1 Protein has been validated for the following applications: Enzyme Activity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a 0.2μm filtered solution in Tris, NaCl, DTT and Glycerol.
Gene ID (Entrez) 23411
Molecular Weight (g/mol) 82 kDa
Name Sirtuin 1/SIRT1
Quantity 50μg
Source E. coli-derived human Sirtuin 1/SIRT1 protein Met1-Ser747,with a C-terminal 6-His tag.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 75%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.