missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SIRT3 Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | SIRT3 Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MSDKGKLSLQ DVAELIRARA CQRVVVMVGA GISTPSGIPD FRSPGSGLYS NLQQYDLPYP EAIFELPFFF HNPKPFFTLA KELYPGNYKP NVTHYFLRLL HDKGLLLRLY TQNIDGLERV SGIPASKLVE AHGTFASATC TVCQRPFPGE DIRADVMADR VPRCPVCTGV VKPDIVFFGE PLPQRFLLHV VDFPMADLLL ILGTSLEVEP FASLTEAVRS SVPRLLINRD LVGPLAWHPR SRDVAQLGDV VHGVESLVEL LGWTEEMRDL VQRETGKLDG PDK |
| Buffer | The SIRT3 protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0) 1mM DTT, 0.1M NaCl, 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction