missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human sIL 2R Protein

A cDNA sequence encoding the sIL 2R was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13503-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933779

  • £155.00 / 25µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

3559
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MDSYLLMWGL LTFIMVPGCQ AELCDDDPPE IPHATFKAMA YKEGTMLNCE CKRGFRRIKS GSLYMLCTGN SSHSSWDNQC QCTSSATRNT TKQVTPQPEE QKERKTTEMQ SPMQPVDQAS LPGHCREPPP WENEATERIY HFVVGQMVYY QCVQGYRALH RGPAESVCKM THGKTRWTQP QLICTGEMET SQFPGEEKPQ ASPEGRPESE TSCLVTTTDF QIQTEMAATM ETSIFTTEYQ VAVAGCVFLL ISVLLLSGLT WQRRQRKSRR TI
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
sIL 2R Protein
Research Use Only
IL2RA
Human
His
IL-2R His-Tag protein is supplied in 1x PBS and 50% glycerol pH 7.4.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers