missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Siglec-3/CD33 Protein, Atto 647N Conjugate
Description
Measured by flow cytometry for its ability to bind anti-Human Siglec-3/CD33 Monoclonal Antibody conjugated fluorescent beads.
Specifications
Specifications
| Accession Number | AAA51948.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 945 |
| Molecular Weight (g/mol) | M.W.-observed: 67-80 kDa, under reducing conditions, M.W.-theoretical: 53.4 kDa (monomer) |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived human Siglec-3/CD33 protein Human Siglec-3/CD33 (Asp18-His259) (N-terminus) Accession # AAA51948.1 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD33, CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, gp67, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, Siglec3, SIGLEC-3, SIGLEC3gp67 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?