missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SHMT1 Protein
A cDNA sequence encoding the SHMT1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13497-1mg
Additional Details : Weight : 0.01000kg
Specifications
SHMT1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MTMPVNGAHK DADLWSSHDK MLAQPLKDSD VEVYNIIKKE SNRQRVGLEL IASENFASRA VLEALGSCLN NKYSEGYPGQ RYYGGTEFID ELETLCQKRA LQAYKLDPQC WGVNVQPYSG SPANFAVYTA LVEPHGRIMG LDLPDGGHLT HGFMTDKKKI SATSIFFESM PYKVNPDTGY INYDQLEENA RLFHPKLIIA GTSCYSRNLE YARLRKIADE NGAYLMADMA HISGLVAAGV VPSPFEHCHV VTTTTHKTLR GCRAGMIFYR KGVKSVDPKT GKEILYNLES LINSAVFPGL QGGPHNHAIA GVAVALKQAM TLEFKVYQHQ VVANCRALSE ALTELGYKIV TGGSDNHLIL VDLRSKGTDG GRAEKVLEAC SIACNKNTCP GDRSALRPSG LRLGTPALTS RGLLEKDFQK VAHFIHRGIE LTLQIQSDTG VRATLKEFKE RLAGDKYQAA VQALREEVES FASFFPLPGL PDF | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SHMT1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 10% glycerol. |