missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SHC1 Protein
A cDNA sequence encoding the SHC1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13493-1mg
Additional Details : Weight : 0.01000kg
Specifications
SHC1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MNKLSGGGGR RTRVEGGQLG GEEWTRHGSF VNKPTRGWLH PNDKVMGPGV SYLVRYMGCV EVLQSMRALD FNTRTQVTRE AISLVCEAVP GAKGATRRRK PCSRPLSSIL GRSNLKFAGM PITLTVSTSS LNLMAADCKQ IIANHHMQSI SFASGGDPDT AEYVAYVAKD PVNQRACHIL ECPEGLAQDV ISTIGQAFEL RFKQYLRNPP KLVTPHDRMA GFDGSAWDEE EEEPPDHQYY NDFPGKEPPL GGVVDMRLRE GAAPGAARPT APNAQTPSHL GATLPVGQPV GGDPEVRKQM PPPPPCPAGR ELFDDPSYVN VQNLDKARQA VGGAGPPNPA INGSAPRDLF DMKPFEDALR VPPPPQSVSM AEQLRGEPWF HGKLSRREAE ALLQLNGDFL VRESTTTPGQ YVLTGLQSGQ PKHLLLVDPE GVVRTKDHRF ESVSHLISYH MDNHLPIISA GSELCLQQPV ERKL | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SHC1 protein (1 mg/ml) is supplied in 20mM Tris-HCl, pH 8.0, 2mM DTT, 200mM NaCl, 0.1mM PMSF and 20% Glycerol. |