missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SFRP5 Protein

A cDNA sequence encoding the SFRP5 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13480-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15943709

  • £155.00 / 20µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SFRP5 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSEEYDYYG WQAEPLHGRS YSKPPQCLDI PADLPLCHTV GYKRMRLPNL LEHESLAEVK QQASSWLPLL AKRCHSDTQV FLCSLFAPVC LDRPIYPCRS LCEAVRAGCA PLMEAYGFPW PEMLHCHKFP LDNDLCIAVQ FGHLPATAPP VTKICAQCEM EHSADGLMEQ MCSSDFVVKM RIKEIKIENG DRKLIGAQKK KKLLKPGPLK RKDTKRLVLH MKNGAGCPCP QLDSLAGSFL VMGRKVDGQL LLMAVYRWDK KNKEMKFAVK FMFSYPCSLY YPFFYGAAEP H
Greater than 85.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SFRP5 protein solution (1 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers