missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Serum Amyloid A1 Protein

Product Code. 18487691 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 18487691

Brand: Novus Biologicals™ NBP23487510ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Fully biologically active when compared to standard. The biological activity determined by a chemoattract bioassay using human monocytes is in a concentration range of 10-100 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P0DJI8, P0DJI8
Concentration LYOPH
For Use With (Application) PAGE
Formulation Lyophilized from a 0.2 um filtered concentrated solution in 20 mM Tris-HCl, pH 9.0, 150 mM NaCl.
Gene ID (Entrez) 6288
Molecular Weight (g/mol) 11.7 kDa
Quantity 10 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Gene Alias MGC111216, PIG4, SAA, SAA2, serum amyloid A protein, serum amyloid A1, TP53I4, tumor protein p53 inducible protein 4
Gene Symbol SAA1
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Research Category Biologically Active Proteins
Purity or Quality Grade >98% pure by SDS-PAGE and HPLC
Protein Serum Amyloid A1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.