missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Serum Amyloid A Protein

Product Code. 15952718
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 15952718

Brand: enQuireBio™ QDX52100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_000322.2
Gene ID (Entrez) 6288
Name Serum Amyloid A Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Untested
Gene Symbol SAA1
Product Type Recombinant Protein
Cross Reactivity Human
Species Human
Protein Tag Native
Buffer See datasheet for buffer information.
Purity or Quality Grade Greater than 95% as verified by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.