missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SerpinB4 Protein
A DNA sequence encoding the Homo sapiens (Human) SerpinB4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6670-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002965.1 | |
SerpinB4 Protein | |
Research Use Only | |
SERPINB4 | |
Recombinant Protein | |
E. coli | |
MNSLSEANTK FMFDLFQQFR KSKENNIFYS PISITSALGM VLLGAKDNTA QQISKVLHFD QVTENTTEKA ATYHVDRSGN VHHQFQKLLT EFNKSTDAYE LKIANKLFGE KTYQFLQEYL DAIKKFYQTS VESTDFANAP EESRKKINSW VESQTNEKIK NLFPDGTIGN DTTLVLVNAI YFKGQWENKF KKENTKEEKF WPNKNTYKSV QMMRQYNSFN FALLEDVQAK VLEIPYKGKD LSMIVLLPNE IDGLQKLEEK LTAEKLMEWT SLQNMRETCV DLHLPRFKME ESYDLKDTLR TMGMVNIFNG DADLSGMTWS HGLSVSKVLH KAFVEVTEEG VEAAAATAVV VVELSSPSTN EEFCCNHPFL FFIRQNKTNS ILFYGRFSSP | |
Greater than 90% as determined by SDS-PAGE. |
6318 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |