missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SerpinB2 protein is fully biologically active when compared to standard. The biologically active was determined by its inhibitory effect against single chain tPA induced cleavage of a chromogenic substrate in Imidazole Buffer at 37 degrees C. Half maximal inhibition against 1.0 μg/mL of single chain tPA was at a concentration of 1.0 μg/mL.
Specifications
Specifications
| Accession Number | P05120, P05120, P05120 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM Tris-HCl, pH 8.0, 150 mM NaCl, 1 mM Cysteine, with . |
| Gene ID (Entrez) | 5055 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 46.6 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/μg of SerpinB2 as determined by LAL method. |
| Gene Alias | HsT1201, Monocyte Arg-serpin, PAI, PAI 2, PAI-2, PAI2Urokinase inhibitor, Placental plasminogen activator inhibitor, PLANH2PAI-2, plasminogen activator inhibitor 2, plasminogen activator inhibitor, type II (arginine-serpin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 2, Serpin B2, serpin peptidase inhibitor, clade B (ovalbumin), member 2 |
| Gene Symbol | SERPINB2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?