missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SERPINA1 Protein

A cDNA sequence encoding the SERPINA1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13455-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933619

  • £2975.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SERPINA1 Protein
Research Use Only
Recombinant Protein
Human
MEDPQGDAAQ KTDTSHHDQD HPTFNKITPN LAEFAFSLYR QLAHQSNSTN IFFSPVSIAT AFAMLSLGTK ADTHDEILEG LNFNLTEIPE AQIHEGFQEL LRTLNQPDSQ LQLTTGNGLF LSEGLKLVDK FLEDVKKLYH SEAFTVNFGD TEEAKKQIND YVEKGTQGKI VDLVKELDRD TVFALVNYIF FKGKWERPFE VKDTEEEDFH VDQVTTVKVP MMKRLGMFNI QHCKKLSSWV LLMKYLGNAT AIFFLPDEGK LQHLENELTH DIITKFLENE DRRSASLHLP KLSITGTYDL KSVLGQLGIT KVFSNGADLS GVTEEAPLKL SKAVHKAVLT IDEKGTEAAG AMFLEAIPMS IPPEVKFNKP FVFLMIDQNT KSPLFMGKVV NPTQK
Greater than 90% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The protein solution contains 20mM Tris-HCl pH 7.5, 1mM DTT, 10% glycerol, and 2mM EDTA.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers