missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Serpin F1/PEDF Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
1mg
20 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
500µg
5µg
Description
PEDF Protein is fully biologically active when compared to standard. The ED50 as determined by its ability to enhance the adhesion of human Saos2 cells to bovine Collagen I coated plate is less than 2 ng/mL, corresponding to a specific activity of >5.0 x 10^5 IU/mg.
Specifications
Specifications
| Accession Number | P36955, P36955, P36955 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, 150 mM NaCl. |
| Gene ID (Entrez) | 5176 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 44.4 kDa |
| Quantity | 20 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of Serpin F1/PEDF as determined by LAL method. |
| Gene Alias | Cell proliferation-inducing gene 35 protein, EPC-1PIG35, PEDFpigment epithelium-derived factor, pigment epithelium derived factor), member 1, proliferation-inducing protein 35, serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, Serpin F1, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 1 |
| Gene Symbol | SERPINF1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction