missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Serpin A1/alpha-1-Antitrypsin Protein
Description
The Recombinant Human Serpin A1/alpha-1-Antitrypsin Protein is derived from NS0. The Recombinant Human Serpin A1/alpha-1-Antitrypsin Protein has been validated for the following applications: Inhibition Activity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in Tris, NaCl and CaCl 2 with Trehalose. |
| Gene ID (Entrez) | 5265 |
| Molecular Weight (g/mol) | 46 kDa |
| Name | Serpin A1/alpha 1-Antitrypsin |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived human Serpin A1/alpha 1-Antitrypsin protein Glu25-Lys418,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Reconstitution | Reconstitute at 100 μg/mL in sterile 50 mM Tris, 10 mM CaCl 2 and 150 mM NaCl, pH 7.5. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?