missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human SENP6 GST (N-Term) Protein

Product Code. 18308299 Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 18308299

Brand: Novus Biologicals™ H00026054Q0110ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, ELISA, PAGE, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 26054
Molecular Weight (g/mol) 37.84 kDa
Quantity 10 μg
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias EC 3.4.22.-, KIAA0797FLJ11887, Sentrin/SUMO-specific protease SENP6, sentrin-specific protease 6, SSP1KIAA0389, SUMO1/sentrin specific peptidase 6,2810017C20Rik, SUMO1/sentrin specific protease 6, SUMO-1-specific protease 1, SUSP1FLJ11355
Gene Symbol SENP6
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein SENP6
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.