missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Semaphorin 4B Fc Chimera Protein

Product Code. 18749636
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18749636 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18749636 Supplier R&D Systems™ Supplier No. 10262S4050
  • ONLINE EXCLUSIVE PRICE
    £736.19
    £655.20 / 50µg
    Save £80.99
    11% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Measured by its ability to enhance neurite outgrowth of E16-E18 rat embryonic cortical neurons. Recombinant Human Semaphorin 4B Fc Chimera , immobilized at 2.5 μg/mL on a 96 well plate, is able to significantly enhance neurite outgrowth.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NPR2.4
For Use With (Application) Bioactivity
Gene ID (Entrez) 10509
Molecular Weight (g/mol) 101 kDa
Name KIAA1745sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4B, MGC131831, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4B, SEMA4B, SEMAC, semaphorin-4B, SemC
Quantity 50 μg
Source

Mouse myeloma cell line, NS0-derived human Semaphorin 4B protein

Human Semaphorin 4B (Leu44-Glu717) Accession # Q9NPR2.4IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.