missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Secretory phospholipase A2 receptor Protein

A DNA sequence encoding the Homo sapiens (Human) Secretory phospholipase A2 receptor, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7606-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049730

  • £1265.00 / 500µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001007268.1
Secretory phospholipase A2 receptor Protein
Research Use Only
PLA2R1
Recombinant Protein
Yeast
EEKTWHEALR SCQADNSALI DITSLAEVEF LVTLLGDENA SETWIGLSSN KIPVSFEWSN DSSVIFTNWH TLEPHIFPNR SQLCVSAEQS EGHWKVKNCE ERLFYICKKA GHVLSDAESG CQEGWERHGG FCYKID
Greater than 90% as determined by SDS-PAGE.
22925
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers