Learn More
enQuireBio™ Recombinant Human Secretin Protein
Recombinant Protein
Brand: enQuireBio™ QP13449-5mg
Additional Details : Weight : 0.01000kg
Specifications
Secretin Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAPRPLLLLL LLLGGSAARP APPRARRHSD GTFTSELSRL REGARLQRLL QGLVGKRSEQ DAENSMAWTR LSAGLLCPSG SNMPILQAWM PLDGTWSPWL PPGPMVSEPA GAAAEGTLRP R | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
5 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The protein (1 mg/ml) was lyophilized with no additives. |