Learn More
enQuireBio™ Recombinant Human Secretin Protein
Recombinant Protein
Brand:  enQuireBio™ QP13449-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15943599
  • £99.15 / 1mg
Estimated Shipment: 12-06-2024
to see stock.
Specifications
Secretin Protein
Research Use Only
Recombinant Protein
E. coli
MAPRPLLLLL LLLGGSAARP APPRARRHSD GTFTSELSRL REGARLQRLL QGLVGKRSEQ DAENSMAWTR LSAGLLCPSG SNMPILQAWM PLDGTWSPWL PPGPMVSEPA GAAAEGTLRP R
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The protein (1 mg/ml) was lyophilized with no additives.