missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SDHA Protein
A DNA sequence encoding the Homo sapiens (Human) SDHA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6659-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001281261.1 | |
SDHA Protein | |
Research Use Only | |
SDHA | |
Recombinant Protein | |
E. coli | |
SAKVSDSISA QYPVVDHEFD AVVVGAGGAG LRAAFGLSEA GFNTACVTKL FPTRSHTVAA QGGINAALGN MEEDNWRWHF YDTVKGSDWL GDQDAIHYMT EQAPAAVVEL ENYGMPFSRT EDGKIYQRAF GGQSLKFGKG GQAHRCCCVA DRTGHSLLHT LYGRSLRYDT SYFVEYFALD LLMENGECRG VIALCIEDGS IHRIRAKNTV VATGGYGRTY FSCTSAHTST GDGTAMITRA GLPCQDLEFV QFHPTGIYGA GCLITEGCRG EGGILINSQG ERFMERYAPV AKDLASRDVV SRSMTLEIRE GRGCGPEKDH VYLQLHHLPP EQLATRLPGI SETAMIFAGV DVTKEPIPVL PTVHYNMGGI PTNYKGQVLR HVNGQDQIVP GLYACGEAAC ASVHGANRLG ANSLLDLVVF GRACALSIEE SCRPGDKVPP IKPNAGEESV MNLDKLRFAD GSIRTSELRL SMQKSMQNHA AVFRVGSVLQ EGCGKISKLY GDLKHLKTFD RGMVWNTDLV ETLELQNLML CALQTIYGAE ARKESRGAHA REDYKVRIDE YDYSKPIQGQ QKKPFEEHWR KHTLSYVDVG TGKVTLEYRP VIDKTLNEAD CATVPPAIRS Y | |
Greater than 80% as determined by SDS-PAGE. |
6389 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |