missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SDF2 Protein
A DNA sequence encoding the Homo sapiens (Human) SDF2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6657-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_008854.2 | |
SDF2 Protein | |
Research Use Only | |
SDF2 | |
Recombinant Protein | |
E. coli | |
SSLGVVTCGS VVKLLNTRHN VRLHSHDVRY GSGSGQQSVT GVTSVDDSNS YWRIRGKSAT VCERGTPIKC GQPIRLTHVN TGRNLHSHHF TSPLSGNQEV SAFGEEGEGD YLDDWTVLCN GPYWVRDGEV RFKHSSTEVL LSVTGEQYGR PISGQKEVHG MAQPSQNNYW KAMEGIFMKP SELLKAEAHH AEL | |
Greater than 90% as determined by SDS-PAGE. |
6388 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |