Learn More
DiscoverX Recombinant Human SCF
Product Code. 17322449
Encompass
Click to view available options
:
2 μg
Product Code. 17322449
Brand: DiscoverX 921114
Recombinant Human SCF is an agonist for c-KIT (CD117). Provided as 2μg of dry powder alongside reconstitution buffer.
Stem cell factor (SCF) is a cytokine with important roles in hematopoiesis, spermatogenesis, and melanogenesis. It potently binds the c-KIT receptor (CD117) and other cytokines to preserve the viability and facilitate the proliferation and differentiation of hematopoietic stem and progenitor cells. Soluble and transmembrane versions of SCF are involved in hematopoesis. Provided as 2μg of dry powder alongside reconstitution buffer.
Specifications
Quantity 2 μg
Product Type Control Ligands