missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SART3 Protein

A DNA sequence encoding the Homo sapiens (Human) SART3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7616-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16079810

  • £467.00 / 100µg
Estimated Shipment: 16-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_055521.1
SART3 Protein
Research Use Only
SART3
Recombinant Protein
Yeast
QRKRARAEKK ALKKKKKIRG PEKRGADEDD EKEWGDDEEE QPSKRRRVEN SIPAAGETQN VEVAAGPAGK CAAVDVEPPS KQKEKAASLK RDMPKVLHDS SKDSITVFVS NLPYSMQEPD TKLRPLFEAC GEVVQIRPIF SNRGDFRGYC YVEFKEEKSA LQALEMDRKS VEGRPMFVSP CVDKSKNPDF KVFRYSTSLE KHKLFISGLP FSCTKEELEE ICKAHGTVKD LRLVTNRAGK PKGLAYVEYE NESQASQAVM KMDGMTIKEN IIKVAISNPP QRKVPEKPET RKAPGGPMLL P
Greater than 90% as determined by SDS-PAGE.
9733
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers