missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sal-like protein 1
A DNA sequence encoding the Homo sapiens (Human) Sal-like protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP10146-ma-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001278375.1 | |
Sal-like protein 1 | |
Research Use Only | |
SALL2 | |
Recombinant Protein | |
Mammal | |
MSRRKQRKPQ QLISDCEGPS ASENGDASEE DHPQVCAKCC AQFTDPTEFL AHQNACSTDP PVMVIIGGQE NPNNSSASSE PRPEGHNNPQ VMDTEHSNPP DSGSSVPTDP TWGPERRGEE SPGHFLVAAT GTAAGGGGGL ILASPKLGAT PLPPESTPAP PPPPPPPPPP GVGSGHLNIP LILEELRVLQ QRQIHQMQ | |
Greater than 90% as determined by SDS-PAGE. |
6297 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
Flag | |
Tris-based buffer, 50% glycerol |