missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Saitohin Protein

A DNA sequence encoding the Homo sapiens (Human) Saitohin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6736-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16075470

  • £384.00 / 100µg
Estimated Shipment: 15-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001007533.1
Saitohin Protein
Research Use Only
STH
Recombinant Protein
E. coli
MSEGGGQVSC IFAAPTRLCR WPALIECGVN LTQPLCEWMI QVARDRTLSL AWEVASLLTL SSSEVGLEGV GTIWPSSYSS EESSRNGAEQ GRQLSIEGPF QGQNCPSHPA AALPLPMRGE SQATSCQV
Greater than 90% as determined by SDS-PAGE.
246744
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers