missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Saitohin Protein
A DNA sequence encoding the Homo sapiens (Human) Saitohin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6736-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001007533.1 | |
Saitohin Protein | |
Research Use Only | |
STH | |
Recombinant Protein | |
E. coli | |
MSEGGGQVSC IFAAPTRLCR WPALIECGVN LTQPLCEWMI QVARDRTLSL AWEVASLLTL SSSEVGLEGV GTIWPSSYSS EESSRNGAEQ GRQLSIEGPF QGQNCPSHPA AALPLPMRGE SQATSCQV | |
Greater than 90% as determined by SDS-PAGE. |
246744 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |