missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human S100A9 Protein

Product Code. 18377727
Change view
Click to view available options
:
Recombinant Human S100A9 Protein, Carrier-Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18377727 Recombinant Human S100A9 Protein, Carrier-Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377727 Supplier R&D Systems™ Supplier No. 9254S9050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human S100A9 Protein is derived from E. coli. The Recombinant Human S100A9 Protein has been validated for the following applications: Bioactivity.

Order Info

The product is shipped at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Source E. coli-derived human S100A9 protein Thr2-Pro114
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias CAGBMigration inhibitory factor-related protein 14, calgranulin B, calgranulin-B, Calprotectin L1H subunit, CFAGMRP-14,60B8AG, CGLB, Human S100A9 Capture, L1AG, LIAG, MAC387, MIF, MRP-14, MRP14Leukocyte L1 complex heavy chain, NIF, P14, protein S100-A9, S100 calcium binding protein A9, S100 calcium binding protein A9 (calgranulin B), S100 calcium-binding protein A9, S100 calcium-binding protein A9 (calgranulin B)
Conjugate Unconjugated

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.