missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human S100A9 Protein

A cDNA sequence encoding the S100A9 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13400-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15943449

  • £155.00 / 25µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

S100A9 Protein
Research Use Only
Recombinant Protein
E. coli
MTCKMSQLER NIETIINTFH QYSVKLGHPD TLNQGEFKEL VRKDLQNFLK KENKNEKVIE HIMEDLDTNA DKQLSFEEFI MLMARLTWAS HEKMHEGDEG PGHHHKPGLG EGTPLEHHHH HH
Greater than 90% as determined by SDS-PAGE.
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
S100A9 Human solution containing 20mM Tris HCl pH 8.0, 0.1M NaCl and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers