missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human S100A8/S100A9 Heterodimer Protein

Product Code. 18365637
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18365637 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18365637 Supplier R&D Systems™ Supplier No. 8226S8050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

The Recombinant Human S100A8/S100A9 Heterodimer Protein is derived from E. coli. The Recombinant Human S100A8/S100A9 Heterodimer Protein has been validated for the following applications: Binding Activity.
TRUSTED_SUSTAINABILITY

Specifications

Concentration >95% by SDS-PAGE w/silver stain
Quantity 50 μg
Source E. coli-derived human S100A8/S100A9 Heterodimer protein Met1-Glu93 (S100A8) & Thr2-Pro114 (S100A9)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.