missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human S100A8 Protein

Product Code. 18785506
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18785506

Brand: R&D Systems™ 9876S8050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human S100A8 Protein is derived from E. coli. The Recombinant Human S100A8 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05109
For Use With (Application) Bioactivity
Gene ID (Entrez) 6279
Molecular Weight (g/mol) 11 kDa
Quantity 50 μg
Source E. coli-derived human S100A8 protein Met1-Glu93
Storage Requirements 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Specific Reactivity S100A8
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 150 μg/mL in water.
Buffer Lyophilized from a 0.2 μm filtered solution in Tris and TCEP with Trehalose.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.