missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100A14 / S114 Protein
A DNA sequence encoding the Homo sapiens (Human) S100A14 / S114, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8145-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_065723.1 | |
S100A14 / S114 Protein | |
Research Use Only | |
S100A14 | |
Recombinant Protein | |
E. coli | |
MGQCRSANAE DAQEFSDVER AIETLIKNFH QYSVEGGKET LTPSELRDLV TQQLPHLMPS NCGLEEKIAN LGSCNDSKLE FRSFWELIGE AAKSVKLERP VRGH | |
Greater than 80% as determined by SDS-PAGE. |
57402 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |