missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human S100A11 Protein

Product Code. 18738567
Click to view available options
:
50μg
This item is not returnable. View return policy

Product Code. 18738567

Brand: R&D Systems™ 9015S11050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human S100A11 Protein is derived from E. coli. The Recombinant Human S100A11 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P31949
For Use With (Application) Bioactivity
Gene ID (Entrez) 6282
Molecular Weight (g/mol) Observed M.W.: 9kDa, reducing conditions, Theoretical M.W.: 12kDa
Quantity 50μg
Source E. coli-derived human S100A11 protein Ala2-Thr105
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias calgizzarin, Metastatic lymph node gene 70 protein, MLN 70, MLN70, protein S100-A11, Protein S100-C, S100 calcium binding protein A11, S100 calcium-binding protein A11, S100 calcium-binding protein A11 (calgizzarin), S100C, S100CS100 calcium binding protein A11 (calgizzarin)
Common Name S100A11
Gene Symbol S100A11
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human ANXA-2 is immobilized at 0.5μg/mL (100μL/well), the concentration of Recombinant Human S100A11 that produces 50% of the optimal binding response is approximately 0.4-2.4μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived Ala2-Thr105
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.