missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human S100A10 Protein

A cDNA sequence encoding the S100A10 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13388-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15943409

  • £5285.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

S100A10 Protein
Research Use Only
Recombinant Protein
E. coli
MPSQMEHAME TMMFTFHKFA GDKGYLTKED LRVLMEKEFP GFLENQKDPL AVDKIMKDLD QCRDGKVGFQ SFFSLIAGLT IACNDYFVVH MKQKGKK
Greater than 95% as determined by SDS-PAGE. 2 bands on Western blot at 15.3 and 30.6 kDa, respectively representing monomeric and dimeric form.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
S100-A10 is supplied in 1xPBS and 50% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers