missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RXRA Protein
A cDNA sequence encoding the RXRA was constructed and used to recombinantly synthesize the protein.
£99.15 - £1985.00
Specifications
Name | RXRA Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933379
|
enQuireBio™
QP13378-10UG |
10 μg |
£99.15
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15953379
|
enQuireBio™
QP13378-50UG |
50 μg |
£155.00
50µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15943379
|
enQuireBio™
QP13378-1MG |
1 mg |
£1985.00
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
RXRA Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The protein (1 mg/ml) containing 20mM Tris-HCl pH 7.5, 0.1M NaCl and 5mM ?-Mercaptoethanol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MLGLNGVLKV PAHPSGNMAS FTKHICAICG DRSSGKHYGV YSCEGCKGFF KRTVRKDLTY TCRDNKDCLI DKRQRNRCQY CRYQKCLAMG MKREAVQEER QRGKDRNENE VESTSSANE | |
Greater than 95.0% as determined by SDS-PAGE. |