missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RRM2 Protein
A DNA sequence encoding the Homo sapiens (Human) RRM2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6630-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001025.1 | |
RRM2 Protein | |
Research Use Only | |
RRM2 | |
Recombinant Protein | |
E. coli | |
MLSLRVPLAP ITDPQQLQLS PLKGLSLVDK ENTPPALSGT RVLASKTARR IFQEPTEPKT KAAAPGVEDE PLLRENPRRF VIFPIEYHDI WQMYKKAEAS FWTAEEVDLS KDIQHWESLK PEERYFISHV LAFFAASDGI VNENLVERFS QEVQITEARC FYGFQIAMEN IHSEMYSLLI DTYIKDPKER EFLFNAIETM PCVKKKADWA LRWIGDKEAT YGERVVAFAA VEGIFFSGSF ASIFWLKKRG LMPGLTFSNE LISRDEGLHC DFACLMFKHL VHKPSEERVR EIIINAVRIE QEFLTEALPV KLIGMNCTLM KQYIEFVADR LMLELGFSKV FRVENPFDFM ENISLEGKTN FFEKRVGEYQ RMGVMSSPTE NSFTLDADF | |
Greater than 90% as determined by SDS-PAGE. |
6241 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |