missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human RPSAP52 GST (N-Term) Protein

Codice prodotto. 18531772 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantity:
10 μg
25 μg
Dimensione della confezione:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
18531772 10 μg 10µg
18409499 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18531772 Fornitore Novus Biologicals™ N. del fornitore H00204010P0110ug

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifica

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 204010
Molecular Weight (g/mol) MolecularWeight-theroretical: 48.2 kDa
Quantity 10 μg
Storage Requirements Store at -80°C. Avoid freeze/thaw cycles.
Gene Alias MGC104447, ribosomal protein SA pseudogene 52, RPSA_17_1251
Gene Symbol RPSAP52
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein RPSAP52
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.