missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RPS5 Protein
A cDNA sequence encoding the RPS5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13359-2ug
Additional Details : Weight : 0.01000kg
Specifications
RPS5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMTEWETA APAVAETPDI KLFGKWSTDD VQINDISLQD YIAVKEKYAK YLPHSAGRYA AKRFRKAQCP IVERLTNSMM MHGRNNGKKL MTVRIVKHAF EIIHLLTGEN PLQVLVNAII NSGPREDSTR IGRAGTVRRQ AVDVSPLRRV NQAIWLLCTG AREAAFRNIK TIAECLADEL INAAKGSSNS YAIKKKDELE RVAKSNR | |
Greater than 90% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RPS5 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.2M NaCl, 50% glycerol and 2mM DTT. |