missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RPS5 Protein

A cDNA sequence encoding the RPS5 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13359-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15963319

  • £155.00 / 10µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

RPS5 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMTEWETA APAVAETPDI KLFGKWSTDD VQINDISLQD YIAVKEKYAK YLPHSAGRYA AKRFRKAQCP IVERLTNSMM MHGRNNGKKL MTVRIVKHAF EIIHLLTGEN PLQVLVNAII NSGPREDSTR IGRAGTVRRQ AVDVSPLRRV NQAIWLLCTG AREAAFRNIK TIAECLADEL INAAKGSSNS YAIKKKDELE RVAKSNR
Greater than 90% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
RPS5 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.2M NaCl, 50% glycerol and 2mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers