missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RPL26L1 Protein

A cDNA sequence encoding the RPL26L1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13346-2ug

 View more versions of this product

Product Code. 15963279

  • £99.15 / 2µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

51121
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMKFNPFV TSDRSKNRKR HFNAPSHVRR KIMSSPLSKE LRQKYNVRSM PIRKDDEVQV VRGHYKGQQI GKVVQVYRKK YVIYIERVQR EKANGTTVHV GIHPSKVVIT RLKLDKDRKK ILERKAKSRQ VGKEKGKYKE ELIEKMQE
Greater than 90% as determined by SDS-PAGE.
RPL26L1 Protein
Research Use Only
RPL26L1
Human
His
The RPL26L1 solutioncontains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 2mM DTT and 40% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers