missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RPE Protein

A cDNA sequence encoding the RPE was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13339-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15943259

  • £155.00 / 20µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

6120
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMASGCK IGPSILNSDL ANLGAECLRM LDSGADYLHL DVMDGHFVPN ITFGHPVVES LRKQLGQDPF FDMHMMVSKP EQWVKPMAVA GANQYTFHLE ATENPGALIK DIRENGMKVG LAIKPGTSVE YLAPWANQID MALVMTVEPG FGGQKFMEDM MPKVHWLRTQ FPSLDIEVDG GVGPDTVHKC AEAGANMIVS GSAIMRSEDP RSVINLLRNV CSEAAQKRSL DR
Greater than 95% as determined by SDS-PAGE.
RPE Protein
Research Use Only
RPE
Human
His
The RPE solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 30% glycerol and 200mM NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers