missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human ROR2 Fc Chimera Protein

Product Code. 18729037
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18729037

Brand: R&D Systems™ 8609RO050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human ROR2 Fc Chimera Protein is derived from NS0. The Recombinant Human ROR2 Fc Chimera Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 4920
Molecular Weight (g/mol) Observed M.W.: 80-92kDa, reducing conditions, Theoretical M.W.: 69kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human ROR2 protein, (N-terminus) Human ROR2 (Val28-Gly403) Accession &Num; AAI30523 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias BDB, BDB1, EC 2.7.10.1, MGC163394, neurotrophic tyrosine kinase receptor-related 2, Neurotrophic tyrosine kinase, receptor-related 2, NTRKR2, NTRKR2tyrosine-protein kinase transmembrane receptor ROR2, receptor tyrosine kinase-like orphan receptor 2
Common Name ROR2
Gene Symbol ROR2
Biological Activity Measured by its binding ability in a functional ELISA. Measured by its ability to bind biotinylated human Wnt-5a in a functional ELISA. The ED50 for this effect is typically 1.5-9μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Human ROR2 (Val28-Gly403) Accession # AAI30523, N-terminus; IEGRMD; Human IgG1 (Pro100-Lys330), C-terminus
Purity or Quality Grade >90%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.