missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ROBLD3 Protein

A cDNA sequence encoding the ROBLD3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13336-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15943249

  • £3965.00 / 1mg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

ROBLD3 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMLRPKA LTQVLSQANT GGVQSTLLLN NEGSLLAYSG YGDTDARVTA AIASNIWAAY DRNGNQAFNE DNLKFILMDC MEGRVAITRV ANLLLCMYAK ETVGFGMLKA KAQALVQYLE EPLTQVAAS
Greater than 95.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The ROBLD3 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M Nacl, 2mM DTT and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers