missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RNF34 Protein
A cDNA sequence encoding the RNF34 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13330-1mg
Additional Details : Weight : 0.01000kg
Specifications
RNF34 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGS MRKAGAT SMWASCCGLL NEVMGTGAVR GQQSAFAGAT GPFRFTPNPE FSTYPPAATE GPNIVCKACG LSFSVFRKKH VCCDCKKDFC SVCSVLQENL RRCSTCHLLQ ETAFQRPQLM RLKVKDLRQY LILRNIPIDT CREKEDLVDL VLCHHGLGSE DDMDTSSLNS SRSQTSSFFT RSFFSNYTAP SATMSSFQGE LMDGDQTSRS GVPAQVQSEI TSANTEDDDD DDDEDDDDEE ENAEDRNPGL SKERVRASLS DLSSLDDVEG MSVRQLKEIL ARNFVNYSGC CEKWELVEKV NRLYKENEEN QKSYGERLQL QDEEDDSLCR ICMDAVIDCV LLECGHMVTC TKCGKRMSEC PICRQYVVRA VHVFKS | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RNF34 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.1M NaCl and 1mM DTT. |