missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RNA-binding protein 1

A DNA sequence encoding the Homo sapiens (Human) RNA-binding protein 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6586-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064340

  • £384.00 / 100µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_006734.1
RNA-binding protein 1
Research Use Only
RBM3
Recombinant Protein
E. coli
MSSEEGKLFV GGLNFNTDEQ ALEDHFSSFG PISEVVVVKD RETQRSRGFG FITFTNPEHA SVAMRAMNGE SLDGRQIRVD HAGKSARGTR GGGFGAHGRG RSYSRGGGDQ GYGSGRYYDS RPGGYGYGYG RSRDYNGRNQ GGYDRYSGGN YRDNYDN
Greater than 90% as determined by SDS-PAGE.
5935
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers