missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ribonucleoprotein PTB-binding 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Ribonucleoprotein PTB-binding 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9902-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_060681.2 | |
Ribonucleoprotein PTB-binding 2 Protein | |
Research Use Only | |
RAVER2 | |
Recombinant Protein | |
Yeast | |
MAAAAGDGGG EGGAGLGSAA GLGPGPGLRG QGPSAEAHEG APDPMPAALH PEEVAARLQR MQRELSNRRK ILVKNLPQDS NCQEVHDLLK DYDLKYCYVD RNKRTAFVTL LNGEQAQNAI QMFHQYSFRG KDLIVQLQPT | |
Greater than 90% as determined by SDS-PAGE. |
55225 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |