missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ribonucleoprotein PTB-binding 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Ribonucleoprotein PTB-binding 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9902-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16018961

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_060681.2
Ribonucleoprotein PTB-binding 2 Protein
Research Use Only
RAVER2
Recombinant Protein
Yeast
MAAAAGDGGG EGGAGLGSAA GLGPGPGLRG QGPSAEAHEG APDPMPAALH PEEVAARLQR MQRELSNRRK ILVKNLPQDS NCQEVHDLLK DYDLKYCYVD RNKRTAFVTL LNGEQAQNAI QMFHQYSFRG KDLIVQLQPT
Greater than 90% as determined by SDS-PAGE.
55225
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers